Anti-EIF3D Rabbit Polyclonal Antibody

Supplier: Proteintech
10219-1-AP-150UL
10086-372EA 709.42 CAD
10086-372
Anti-EIF3D Rabbit Polyclonal Antibody
Antibodies
The mammalian translation initiation factor 3 (eIF3), is a multiprotein complex of ~600 kDa that binds to the 40 S ribosome and promotes the binding of methionyl-tRNAi and mRNA. The EIF3S7(p66) is the major RNA binding subunit in this complex. Human eIF3-p66 shares 64% sequence identity with a hypothetical Caenorhabditis elegans protein, presumably the p66 homolog. Deletion analyses of recombinant derivatives of eIF3-p66 show that the RNA-binding domain lies within an N-terminal 71-amino acid region rich in lysine and arginine.

Western Blot: Mouse Brain Tissue, 1:200-1:1000; IHC: HumanMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol EIF3D
Environmentally Preferable
Antigen synonyms eIF3-p66|EIF3S7|eIF3-zeta
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Immunogen Recombinant Protein
Purification Antigen affinity Purification
Storage buffer PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
Shipping temperature Store at -20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR