Anti-EID1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11734-1-AP-150UL
10086-330EA 709.42 CAD
10086-330
Anti-EID1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: EID1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR