Anti-CSPG4,NG2 Rabbit Polyclonal Antibody

Supplier: Proteintech
55027-1-AP-150UL
10085-260EA 709.42 CAD
10085-260
Anti-CSPG4,NG2 Rabbit Polyclonal Antibody
Antibodies
CSPG4, also named as HMW-MAA, MCSP, MCSPG, MEL-CSPG, MSK16 and NG2, is a proteoglycan playing a role in cell proliferation and migration which stimulates endothelial cells motility during microvascular morphogenesis. CSPG4 may inhibit neurite outgrowth and growth cone collapse during axon regeneration. It is cell surface receptor for collagen alpha 2(VI) which may confer cells ability to migrate on that substrate. CSPG4 may regulate MPP16-dependent degradation and invasion of type I collagen participating in melanoma cells invasion properties. It modulates the plasminogen system by enhancingMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CSPG4
Antigen name Chondroitin Sulfate Proteoglycan 4
Environmentally Preferable
Antigen synonyms MCSP|NG2
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Immunogen Peptide
Purification Antigen affinity Purification
Storage buffer 0.1M NaHCO3, 0.1M glycine, 0.02% sodium azide and 50% glycerol pH 7.3.
Shipping temperature Store at -20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR