Anti-Casein Kinase 1 epsilon Rabbit Polyclonal Antibody

Supplier: Proteintech
11230-1-AP-150UL
10085-252EA 709.42 CAD
10085-252
Anti-Casein Kinase 1 epsilon Rabbit Polyclonal Antibody
Antibodies
CSNK1E(Casein kinase I isoform epsilon) is also named as CKIe, CKI-epsilon and belongs to the casein kinase I subfamily. It plays a significant role in delaying the negative feedback signal within the transcription-translation-based autoregulatory loop that composes the core of the circadian mechanism and makes an ideal target for pharmaceutical compounds influencing circadian rhythms, sleep, and jet lag, as well as other physiologic and metabolic processes under circadian regulation. It can be autophosphorylated.

IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: [More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name Casein Kinase 1 epsilon
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
ELISA Yes
ImmunoChemistry Yes
Immunogen Recombinant Protein
Purification Antigen affinity Purification
Storage buffer PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
Shipping temperature Store at -20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR