Anti-CDH23 Rabbit Polyclonal Antibody

Supplier: Proteintech
13496-1-AP-150UL
10084-510EA 709.42 CAD
10084-510
Anti-CDH23 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A431 Cells, 1:200-1:1000

Type: Primary
Antigen: CDH23
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR