Anti-C10ORF67 Rabbit Polyclonal Antibody
Supplier: Proteintech
24459-1-AP-150UL
10083-746EA 709.42 CAD
10083-746
Anti-C10ORF67 Rabbit Polyclonal Antibody
Antibodies
RNF17 also termed as ring finger protein 17 is a 1623 amino acid protein, which contains 1 ring type zinc finger and 4 tudor domains. RNF17 is specifically expressed in testis. The RING finger motif presents in many ubiquitin E3 ligases. RNF17 interacts with all four members of the Mad family (Mad1, Mxi1, Mad3 and Mad4), which are basic-helix-loop-helix-leucine zipper transcription factors of the Myc oncoprotein network. RNF17 is component of the tectonic-like complex, which is required for ciliogenesis and sonic hedgehog/SHH signaling. RNF17 is able to form dimers or polymers both in vitroMore Product Information