Anti-BTN2A1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20596-1-AP-150UL
10083-722EA 709.42 CAD
10083-722
Anti-BTN2A1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:500-1:5000

Type: Primary
Antigen: BTN2A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR