Anti-ACSL4/FACL4 Rabbit Polyclonal Antibody

Supplier: Proteintech
22401-1-AP-150UL
10082-460EA 709.42 CAD
10082-460
Anti-ACSL4/FACL4 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human Placenta Tissue, 1:20-1:200

Type: Primary
Antigen: ACSL4/FACL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR