Anti-PPP1CC Rabbit Polyclonal Antibody

Supplier: Proteintech
11082-1-AP-150UL
10082-190EA 709.42 CAD
10082-190
Anti-PPP1CC Rabbit Polyclonal Antibody
Antibodies
PPP1CC, also named as PP-1G, belongs to the PPP phosphatase family and PP-1 subfamily. Protein phosphatase 1 (PP1) is essential for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis. It is involved in regulation of ionic conductances and long-term synaptic plasticity. PPP1CC may play an important role in dephosphorylating substrates such as the postsynaptic density-associated Ca2+/calmodulin dependent protein kinase II.

Western Blot: MCF7 Cells, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
More Product Information
Order Now

SPECIFICATIONS

Antigen symbol PPP1CC
Antigen name Serine/threonine-protein phosphatase PP1-gamma catalytic subunit
Environmentally Preferable
Antigen synonyms PP1C|PP-1G|PPP1G
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Immunogen Recombinant Protein
Purification Antigen affinity Purification
Storage buffer PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
Shipping temperature Store at -20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR