Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: Columbia Biosciences
D3-110
10065-774EA 270.27 CAD
10065-774
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR