Anti-MASMTGGQQMG Rabbit Polyclonal Antibody (DyLight® 488)

Supplier: Columbia Biosciences
D8-1832
10065-720EA 865.24 CAD
10065-720
Anti-MASMTGGQQMG Rabbit Polyclonal Antibody (DyLight® 488)
Antibodies
Type:
Antigen: MASMTGGQQMG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR