Anti-GKPIPNPLLGLDST Rabbit Polyclonal Antibody (DyLight® 650)

Supplier: Columbia Biosciences
D10-1834
10065-506EA 890.5 CAD
10065-506
Anti-GKPIPNPLLGLDST Rabbit Polyclonal Antibody (DyLight® 650)
Antibodies
Type:
Antigen: GKPIPNPLLGLDST
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR