Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Columbia Biosciences
AP-112
10065-494EA 402.81 CAD
10065-494
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen symbol IgG
Antigen name Immunoglobulin G
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation AP (Alkaline Phosphatase)
Reactivity Mouse
Host Goat
Isotype IgG
Western blot Yes
ELISA Yes
Storage temperature Store at 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR