Anti-6xHis Mouse Polyclonal Antibody (Biotin) [clone: AD1.1.1]

Supplier: Columbia Biosciences
B1-1711
10065-462EA 901.33 CAD
10065-462
Anti-6xHis Mouse Polyclonal Antibody (Biotin) [clone: AD1.1.1]
Antibodies
Type:
Antigen: 6xHis
Clonality: Polyclonal
Clone: AD1.1.1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR