7000 Series Reusable Half Mask Respirators, Pre-Assembled, Moldex®
Supplier: Moldex/Metric
7111 7112 7113
10065-332BG 91.89 CAD
10065-332 10065-334 10065-336 10065-338 10065-340 10065-342
7000 Series Reusable Half Mask Respirators, Pre-Assembled, Moldex®
Respirators Reusable Respirators Half Face Respirators
Oil-free or Oil/water based paint spray or pesticides pre-assembled respirators.
  • 100% PVC-free
  • NIOSH approved for organic vapors
  • Oil-free or oil/water based
Areas of applications include degreasing, forging, foundries, heat treating operations, paper making, rendering, rubber manufacturing, manufacture and use of adhesives, dry cleaning, pesticide manufacture and application, treatment of timber and wood products, manufacture and use of inks, oil-based paint, printing, manufacture of cosmetic preparations, perfumes and pharmaceuticals, flour bleaching,More Product Information