Quatro Cables With Conductivity Sensors, YSI
Supplier: YSI
605790-1 605790-4 605790-10 605790-20 605790-30
10064-822EA 4130.39 CAD
10064-822 10064-824 10064-826 10064-828 10064-830
Quatro Cables With Conductivity Sensors, YSI
Cables
The Quatro is a new 4-port cable and bulkhead for the Professional Plus handheld instrument that allows users to measure conductivity and temperature, dissolved oxygen, and a choice of any two ISE’s among pH, ORP, ammonium, nitrate, or chloride.
  • Holds four user replaceable Professional Series sensors
  • Choose either polarographic or galvanic DO sensors
  • Weighted guard ensures correct depth while protecting the sensors
  • 2-year cable warranty
  • Cable management kit included on any cable 4 meters or longer
The Professional Plus with the Quatro 4-port cableMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable