VWR® Cell Culture Flasks
Supplier: VWR International
10062-860 10062-862 10062-864 10062-866 10062-868 10062-870 10062-872 10062-874 10062-884 10062-886
10062-860CS 584.86 CAD
10062-860 10062-862 10062-864 10062-866 10062-868 10062-870 10062-872 10062-874 10062-884 10062-886
VWR® Cell Culture Flasks
Flasks Cell Culture Flasks
VWR® Cell culture flasks are vacuum gas plasma treated for consistent cell attachment and growth.
  • Made of crystal clear virgin polystyrene
  • Sterile and certified non-pyrogenic
  • Volume graduations on both sides with special writing area
  • Choice of vented or plug seal caps
The angled-neck design of these flasks allows access to the entire growth surface with either pipets or cell scrapers. Flasks have volume graduations on both sides, and a writing surface to facilitate sample identification.
Vented caps incorporate a 0.22 µm hydrophobic filter to allowMore Product Information