Ultra Cyano HPLC Prep Columns, Restek
Supplier: Restek
9106577
10059-668EA 3697.93 CAD
10059-668
Ultra Cyano HPLC Prep Columns, Restek
Chromatography Columns
Restek makes it easy to scale up your separations.
  • Easy scale-up from Restek analytical columns
  • Popular bonded phases
We offer a wide range of semipreparative and preparative-scale columns packed with many of our popular Ultra, Allure®, Pinnacle® DB, and Viva bonded phases on a 5 μm spherical silica.
The Restek® Cyano is a traditional monomeric cyanopropylsilane that is recommended for assays where alternate selectivity, or confirmation, to a C18 or C8 column is desired. It can be used in normal-phase, reversed-phase (best with mobile phase pHMore Product Information
Order Now
SPECIFICATIONS
Description 10059-668
Environmentally Preferable
Carbon load Carbon Load: 8%
Endcapped Yes
pH range 2.5 to 8
Temperature limit 80°C