Pinnacle® DB PFP Propyl HPLC Columns (USP L43), Restek
Supplier: Restek
9419332 9419352 9419355 9419362 9419365 9419512 9419515 9419525 9419552 9419562 9419565 9419572 9419575 9419312 9419315 941931E 941933E 941936E 941956E 941957E 9419212 9419232 9419252 9419335 9419522 9419532 9419535 9419555 941935E 941951E 941952E 941953E 941955E
10058-104EA 984.64 CAD
10058-104 10058-108 10058-110 10058-112 10058-114 10058-116 10058-118 10058-120 10058-122 10058-124 10058-126 10058-128 10058-130 10059-910 10059-912 10060-136 10060-138 10060-140 10060-142 10060-144 10839-944 10839-946 10839-948 10839-954 10839-968 10839-972 10839-974 10839-978 10841-090 10841-092 10841-094 10841-096 10841-098 10841-114 10841-116 10841-118 10841-122 10841-124 10841-126 10841-128 10841-130 10841-132 10841-134 10841-136 10841-138 10841-140 10841-142 10841-144 10841-148 10841-152 10841-154 10841-156 10841-158 10841-160 10841-162 10841-164 10841-166 10841-168 10841-170 10841-172 10841-174 10841-176 10841-178
Pinnacle® DB PFP Propyl HPLC Columns (USP L43), Restek
Chromatography Columns
The Restek PFP Propyl is a great choice for the retention and selectivity of charged bases, electronegative compounds, and amine-containing compounds.
  • For retention and selectivity of charged bases
  • More amenable to LC-MS
  • Efficient with acidic mobile phases
Unlike a conventional cyano column, the Restek PFP Propyl is much more amenable to LC-MS because it is more reliable and efficient with acidic mobile phases.
Pinnacle DB columns are 100% manufactured by Restek in our Bellefonte, Pennsylvania facility. Because performance begins with the support,More Product Information