LC OQ Linearity Test Mix Kit, Restek
Supplier: RESTEK CORP
31805
10058-056KT 325.46 CAD
10058-056
LC OQ Linearity Test Mix Kit, Restek
Standards Chromatography Standards
Contains:
  • Caffeine at 5, 25, 125, 250, 500 µg/mL in water in a five-ampul kit.
  • Simple check of a detector’s ability to produce a linear response to varied concentrations.
  • Suitable for clear indication of linear range.