RT Touch Magnetic Stirrers, 230 V
Supplier: Thermo Fisher Scientific
88880014 88880016
10054-190EA 734.04 CAD
10054-190 10054-194
RT Touch Magnetic Stirrers, 230 V
Stirrers Magnetic Stirrers
  • 230 V model
  • Precise Digital Control Capable of Slow Speed Stirring
  • Timer Runs Continuous or Countdown Cycles
  • Low-Profile, Lightweight Design with Small Footprint
  • Strong Magnetic Coupling Ensures Consistent Stirring during Experimental Procedures
  • Brushless DC Motor will not Generate Heat and 'Cook' Samples
  • Polypropylene Top and Body Provide Excellent Chemical Resistance
  • Two Plate Sizes Available: 170 mm, or 220 mm
  • Detachable plug
  • Two Non-Slip Silicone Plate Covers Included (1 Black, 1 White)
  • Gradual Start-Up Stirring to Prevent Splashing
  • Threaded Hole for
More Product Information