RT Basic Series Magnetic Stirrers, Thermo Scientific
Supplier: Thermo Fisher Scientific
88880007 88880009 88880011
10054-162EA 396.86 CAD
10054-162 10054-180 10054-184
RT Basic Series Magnetic Stirrers, Thermo Scientific
Stirrers Magnetic Stirrers
Experience reliability with the plug-and-play RT Basic series magnetic stirrers, which are ideal for routine stirring applications.
  • Low-profile, lightweight design with small footprint
  • Strong magnetic coupling ensures consistent stirring during experimental procedures
  • Brushless DC motor will not generate heat and “cook” samples
  • Polypropylene top and body provides excellent chemical resistance
  • Detachable plug
  • Low power consumption
  • 2-year warranty
Three size options (120, 170, or 220 mm) are available to choose from, all with a low-profile, lightweightMore Product Information