Anti-CD3E Mouse Monoclonal Antibody [clone: OKT3]
Supplier: Tonbo Biosciences
40-0037-U100 40-0037-U500
10051-060EA 165.9 CAD
10051-060 10051-062
Anti-CD3E Mouse Monoclonal Antibody [clone: OKT3]
Antibodies
The OKT3 antibody is specific for human CD3e, also known as CD3 epsilon, a 20 kDa subunit of the T cell receptor complex along with CD3 gamma and CD3 delta. These integral membrane protein chains assemble with additional chains of the T cell receptor (TCR), as well as CD3 zeta chain, to form the T cell receptor – CD3 complex. Together with co-receptors CD4 or CD8, the complex serves to recognize antigens bound to MHC molecules on antigen-presenting cells. These interactions promote T cell receptor signaling (T cell activation), inducing a number of cellular responses including proliferation,More Product Information