Anti-CD3E Armenian Hamster Monoclonal Antibody [clone: 145-2C11]
Supplier: Tonbo Biosciences
40-0031-M001 40-0031-U100 40-0031-U500
10051-050EA 423.44 CAD
10051-050 10051-052 10051-054
Anti-CD3E Armenian Hamster Monoclonal Antibody [clone: 145-2C11]
Antibodies
The 145-2C11 antibody is specific for mouse CD3e, also known as CD3 epsilon, a 20 kDa subunit of the T cell receptor complex, along with CD3 gamma and CD3 delta. These integral membrane protein chains assemble with additional chains of the T cell receptor (TCR), as well as CD3 zeta chain, to form the T cell receptor – CD3 complex. Together with co-receptors CD4 or CD8, the complex serves to recognize antigens bound to MHC molecules on antigen-presenting cells. Such interactions promote T cell receptor signaling (T cell activation) and can result in a number of cellular responses includingMore Product Information