Glutaraldehyde 50% aqueous solution Biological Grade

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial

16500 16501 16521 16520
100504-008EA 85.59 CAD
100504-008 100504-010 100504-012 100505-022
Glutaraldehyde 50% aqueous solution Biological Grade
Glutaraldehyde

  • Highest purity, free from polymers and other contaminants
  • Each lot is tested and assayed in our lab and we guarantee its purity
  • Sealed under dry nitrogen
  • Refrigerated prior to shipment

Formula: C₅H₈O₂
MW: 100.12
Boiling Pt: 188
Melting Pt: -10
Density: 1.014
MDL Number: MFCD00007025
CAS Number: 111-30-8
UN: 3265
ADR: 8 III
Merck Index: 13,04485

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR