Tri-Pour Beaker Caps, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60975 60979 60973 60971 60977 60969
100503-214EA 254.63 CAD
100503-214 100503-222 100503-210 100503-280 100503-218 100503-204
Tri-Pour Beaker Caps, Electron Microscopy Sciences
Closures Bottle Closures
The caps are wax impregnated, have lift-tabs, and may be written on for sample identification.
The caps are available for all Tri-Pour beakers.