Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
11774-05 11774-10 11773-10 11772-05 11773-05 11772-10
100496-434EA 87.91 CAD
100496-434 100503-018 100503-016 100496-430 100496-432 100503-014
Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences
Buffers Biological Buffers
All of our Dullbecco’s phosphate buffer saline solutions contain: potassium chloride, potassium phosphate monobasic, sodium chloride, sodium phosphate dibasic, and D-glucose.
  • Some formulations may contain additional components
  • Store at room temperature
Use the table below as a quick reference for identifying the one that best suits your application. All of the solutions are prepared 1X strength.