Micro-Tool Individual Micro Tools, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62090-00 62091-01-12 62091-24-50 62091-26-25 62091-04-50 62091-25-50 62091-10 62091-18-50 62091-01-50 62091-01-25 62091-11-25 62091-13-0.12 62091-27-50 62091-29-25 62091-31 62091-03-50 62091-05-50 62091-22 62091-11-50 62091-05-25 62091-25-12 62091-27-12 62091-07-0.50 62091-12-25 62091-24-12 62091-26-12 62091-24-25 62091-13-50 62091-15-25 62091-19 62091-27-25 62091-28-25 62091-26-50 62091-08-25 62091-16-0.50 62091-32 62091-28-50 62091-02-50 62091-04-12 62091-06 62091-15-50 62091-25-25 62091-08-50 62091-09-50 62091-04-025 62091-20-25 62091-20-50
100497-626EA 75.37 CAD
100497-626 100497-632 100497-698 100497-708 100501-632 100501-658 100497-664 100501-648 100497-628 100497-630 100497-666 100497-672 100497-712 100497-726 100497-730 100501-628 100501-636 100501-654 100502-470 100497-648 100497-706 100497-716 100497-652 100497-668 100497-700 100497-710 100501-656 100501-644 100497-678 100497-686 100497-714 100497-718 100501-660 100497-654 100497-680 100497-732 100501-662 100497-636 100497-642 100497-646 100497-650 100497-676 100497-704 100501-638 100501-642 102093-924 102093-932 102093-934 102093-938
Micro-Tool Individual Micro Tools, Electron Microscopy Sciences
Tool Sets
Micro-Tools are the smallest known precision tools available for the laboratory and micro electronics industry. These tools are designed for anyone who is engaged in microscope work, miniature applications as well as specimen manipulation.
  • Consists of a threaded handle (MT1), 4.75" long and an interchangeable threaded base with tool tip
  • Available in 32 different tip configurations with interchangeable handles
  • Tip material is hardened tool steel unless noted
Order Now
SPECIFICATIONS
Environmentally Preferable