Graduated Pipettes, Polypropylene, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70952-10 70952-50 70951-02 70952-01 70952-05 70952-25 70952-02
100489-204EA 323.74 CAD
100489-204 100489-208 100489-196 100489-200 100489-202 100489-206 100501-216
Graduated Pipettes, Polypropylene, Electron Microscopy Sciences
Pipettes Graduated Pipettes
Made from translucent polypropylene.
  • Resistant to breakage
  • Blue graduations, ring mark and inscriptions
Ideal for fluoro and silicate chemistry used. Recommended cleaning with mild alkaline detergent up to 60 °C.
NOTE: Exposure to temperatures above 60 °C may cause volume changes.
Order Now
SPECIFICATIONS
Environmentally Preferable