Netwell™ Tissue Processing Well, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64736-01 64742-00 64710-00 64712-00 64735-00 64740-00 64745-00 64713-00 64730-00 64732-00 64732-10 64730-10 64743-00 64736-00 64735-01 64711-00
100498-854EA 139.39 CAD
100498-854 100498-858 100500-454 100500-458 100500-462 100498-856 100498-862 100500-460 100502-620 100502-622 100490-642 100490-640 100498-860 100500-464 100498-852 100500-456
Netwell™ Tissue Processing Well, Electron Microscopy Sciences
Culture Dishes Bioassay Dishes
Home made devices are no longer necessary
These tissue processing wells are ready to fill all of your needs with the added convenience of being able to process up to 12 tissue specimens at one time (with one unit) . Tissue processing well is a modular and disposable system which can be used for the following procedures:
The handling and processing of tissue slices.
In immunocytochemical staining.
In 3-dimensional tissue culture.
In explant tissue culture.
Home made devices are no longer necessary. These tissue processing wells are ready to fill all of your needs with the added convenience ofMore Product Information