British Standard Graticules - NG10, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68026-16 68026-19
100499-242EA 687.57 CAD
100499-242 100499-244
British Standard Graticules - NG10, Electron Microscopy Sciences
Graticules, Reticles and Grids
Originally designed by the National Coal Board for use in coal mining.
In this graticule the circle areas double progressively, hence the diameters alter by Root 2, so that the size classes can form a continuation of the standard series of sieves for particle sizing. Each particle is assigned to a size class defined by two adjacent circles, which represent the size limits of that class. Thus the distribution of size is obtained in terms of the diameter of circles having the same projected area as the particles. This method will cover particles in the range 150 micron to 0.38 micron. TheMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable