Whipple Grid Graticules, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68009-16 68009-19 68009-20.4 68009-23 68009-24.5 68009-25 68009-26 68009-27
100499-128EA 349.59 CAD
100499-128 100499-130 103305-428 103305-430 103305-432 103305-434 103305-436 103305-438
Whipple Grid Graticules, Electron Microscopy Sciences
Graticules, Reticles and Grids
Whipple grid graticules are used for drinking water particle analysis and for other particle counting.
  • Whipple grid 100 squares in 7 mm area
  • Surface chrome image
Grid showed ratio of full square to smallest is 50:1 and area is 2500:1.