Master Coiler Plier, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62186-20
100498-002EA 41.16 CAD
100498-002
Master Coiler Plier, Electron Microscopy Sciences
Pliers
This ʽMaster Coiler’ is the painless plier. It is designed to make loops and coils without nicking precious wire.
  • Comfort grips
  • Non-rust
It allows one to hold the wire with the flat jaw while rotating the round jaw to form the perfect loop.