Formalde-Fresh solution for histology low odor

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

15678-01
100496-490EA 288.46 CAD
100496-490
Formalde-Fresh solution for histology low odor
Formaldehyde

A specially formulated odor-mask which neutralizes the formalin odor without affecting subsequent tissue processing, handling, or staining.


  • Low odor
  • Ready-to-use
  • 10% formalin solution


A specially formulated odor-mask which neutralizes the formalin odor without affecting subsequent tissue processing, handling, or staining. Low odor, ready-to-use 10% formalin solution. May be used as a fixative or preservative of specimens. Its shelf life is 24 months.

Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: -19.3
Melting Pt: -92
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
Merck Index: 13,04259

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR