MAC X-ray Microanalysis Individual Standards, Grains, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
80075-HF 80075-PD 80075-SB 80075-SE 80075-Y 80075-AS 80075-BE 80075-C 80075-PT 80075-TE 80075-CU 80075-NB 80075-RH 80075-SC 80075-AG 80075-AL 80075-MN 80075-SI 80075-CO 80075-GE 80075-SN 80075-CD 80075-RU 80075-TL 80075-ZR 80075-FE 80075-MG 80075-TA 80075-TI 80075-BAS 80075-BAS-FC
100495-174EA 749.6 CAD
100495-174 100495-200 100495-210 100495-214 100495-232 100495-140 100495-154 100495-158 100495-202 100495-222 100495-166 100495-184 100495-206 100495-212 100495-136 100495-138 100495-180 100495-216 100495-162 100495-172 100495-218 100495-160 100495-208 100495-226 100495-236 100495-168 100495-178 100495-220 100495-224 102099-030 102099-032
MAC X-ray Microanalysis Individual Standards, Grains, Electron Microscopy Sciences
Standards Colour Reference and Spectroscopy Standards
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems.
All the standards are polished to a 0.25 micron diamond finish and carbon coated. They are supplied with a location map for standard identification. Grains of standard materials are available and they measure approximately 2 x 2 mm in size. They are supplied with fully authenticated certificates of analysis.
Order Now
SPECIFICATIONS
Environmentally Preferable