EMS Syntheric Fiber Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78460-710 78460-707 78460-708 78460-06 78460-705 78460-702A 78460-709
100494-896EA 18.41 CAD
100494-896 100494-890 100494-892 100494-884 100494-888 100494-886 100494-894
EMS Syntheric Fiber Tweezers, Electron Microscopy Sciences
Forceps
These fiber tweezers are made from SV plastic which is based on polyvinylidene fluoride (PVDF). Very hard, high tensile and flexural strength. Resistant to high heat and thermal aging. Excellent chemical resistance. ESD safe – low surface resistivity.
  • Good mechanical strength and toughness
  • Smooth surface
  • Continuous use temperature up to 150°C
  • High purity (clean room and medical devices approved, low extraction value)
  • Excellent chemical resistance to most aggressive substances
  • Outstanding resistance to HF, HNO3 , HCl (all concentrations. 90°C)
  • Resistant to
More Product Information