Colored Microscope Slide Boxes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71458-W 71459-R 71458-B 71458-R 71459-B 71459-W
100493-348EA 24.79 CAD
100493-348 100493-352 100493-344 100493-346 100493-350 100493-354
Colored Microscope Slide Boxes, Electron Microscopy Sciences
Microscope Slide Boxes
Molded from one piece (lid and bottom) polypropylene these slide boxes are able to accommodate either 25 or 100 standard 75 x 25 mm (3 x 1") slides.
The attached lid is held securely by a closed latch lock. Stacking feet maximize the stability when boxes are stacked up one atop the other. The boxes are unlined and the index number is printed on the bottom. Comes complete with a numbered slide index card. These boxes are available in blue, red or white.