PEAK Achromatic Magnifier; 4X, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68128-04
100492-720EA 301.46 CAD
100492-720
PEAK Achromatic Magnifier; 4X, Electron Microscopy Sciences
Magnifiers
Uses high-resolution achromatic lenses whose chromatic aberration has been corrected.
With its wide square Field-of-View (45x45mm) and its interchangeable semi-transparent/ black skirt, this unique magnifier offers you a variety of uses for precision work. Fine focus adjustment with multi-groove screw helicoid system gives you very smooth operation. Neck strap is included.