Diamond Dissecting Knives, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72027 72025 72026 72029 72032 72030 72031
100491-900EA 3150.3 CAD
100491-900 100491-896 100491-898 100491-904 100491-910 100491-906 100491-908
Diamond Dissecting Knives, Electron Microscopy Sciences
Knives
DIATOME has now begun to manufacture diamond knives and blades for microsurgery, dissection, and its related applications.
  • Ultra-sharp blade cuts with minimum pressure, without tearing or damaging the specimen and reduces the risk of specimen distortion
  • Unique pressure mechanism allows blade actuation with one hand
  • The handle is made of titanium and is coated by high vacuum with titanium nitride to harden the surface.
  • The knife is corrosion free and is completely resistant to saline solutions.
  • It can be sterilized in an autoclave up to 200°C.
  • Special tray
More Product Information