Colormark ™; Plus Glass Slide, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63400-30 63400-41 63400-61 63400-20 63400-40 63400-10 63400-50 63400-31 63400-60
100491-378EA 233.9 CAD
100491-378 100498-460 100491-386 100491-376 100491-380 100491-374 100491-382 100498-458 100491-384
Colormark ™; Plus Glass Slide, Electron Microscopy Sciences
Slides Microscope Slides
Specimen slide identification is vital automated etched slide labeling instruments ensure specimen slide integrity but are difficult to read on ordinary slides.
To clearly read diamond scribed slide labels against any surface, you need the only slide designed to be used with your automated slide labeling instrument.
You need COLORMARK slides.These brightly printed slides greatly improve etched labeling visibility and offer all the advantages of Superfrost slides. A bright contrasting color has been printed on the back of the Superfrost end so that patient or specimen information etchedMore Product Information