BEEM™ Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71148-10 71148-05 71148-01
100490-976EA 1369.4 CAD
100490-976 100490-974 100493-136
BEEM™ Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Boxes Storage Boxes
A two tone color coded plastic box with insert which has 24 letter-labeled crossed slots, where the grids can be stored. A rotating protection plate covers the slots and allows for exposure of one grid at a time. Measures L x W x H: 57 x 45 x 12.5 mm (2 1/3 x 1 3/4 x 1/2").
*BEEM® Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc.