Glass Knife Strips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71014 71015 71013 71012 71016 71018
100489-842EA 818.09 CAD
100489-842 100489-844 100489-840 100489-838 100489-846 100489-848
Glass Knife Strips, Electron Microscopy Sciences
Microtomes and Microtome Assemblies Microtome Blades and Handles
Specially selected glass strips for making glass knives.
Its hardness produces a durable and sharp edge suitable for thick, semi thin, or ultrathin sections.
Order Now
SPECIFICATIONS
Environmentally Preferable