Scintillation Vials, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72632-PT 72634-PT 72632 72634 72632-PE 72634-PE
100489-516EA 190.33 CAD
100489-516 100489-520 100500-676 100500-678 100489-514 100489-518
Scintillation Vials, Electron Microscopy Sciences
Vials
Vials are available in Wheaton 180® low potassium borosilicate glass, High Density Polyethylene (HDPE) and Polyethylene Terephthalate (PET).
  • Screw caps size 22-400 is made from Urea with Poly-seal cone liner
  • Vial measures 28 mm diameter × 57 mm height (with cap off), and 61 mm height (with cap on)
Opaque, high-density polyethylene vials are economically priced. They are 20 ml in capacity and they measure 27 mm diameter × 61 mm height (with cap). The screw caps are 22-400 and they are made from polypropylene with a foam- polyethylene liner.