Muscovite Mica Sheets, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71850-11 71851-05 71857-15 71858-01 71857-01 71854-15 71850-01 71855-05 71853-01 71855-15 71857-11 71853-15 71855-10 71850-15 71856-01 71857-10 71853-11 71855-01 71855-10-10 71855-11-10 71857-15-10 71856-03 71855-15-10 71857-10-10 71856-04 71856-01-10 71856-02 71855-05-10
100488-346EA 27.97 CAD
100488-346 100489-370 100493-522 100499-576 100502-194 100489-384 100501-900 100493-506 100489-374 100493-512 100493-520 100489-382 100493-508 100489-368 100493-514 100493-518 100489-380 100502-192 103301-954 103301-956 103302-312 103302-314 103302-316 103302-318 103302-346 103302-472 103302-494 103302-552
Muscovite Mica Sheets, Electron Microscopy Sciences
Specimen Mounting Supplies
EMS Mica Sheets offer a clean surface for EM applications, carbon filming and particle spraying, as well as for AFM applications. The highest quality mica (V-1 or V-2) for AFM applications to the Medium Quality (V-4 to V-6) for replication and thin film deposition.
  • High Quality Muscovite Mica
  • EM applications, carbon filming and particle spraying and AFM applications
  • Peels off very thin up to 0 0001" uniform layers
There are two types of mica: muscovite and phlogopite. Generally, one differs from the other by color (Muscovite is Ruby, Green or White;More Product Information