Lab-Tek® II Chamber Slide™ System, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70360-26 70360-24 70360-46 70379-16 70360-12 70360-82 70379-44 70379-84 70379-46 70360-22 70379-86 70379-42 70360-14 70379-22 70379-26 70360-42 70360-44 70379-12 70379-82
100489-082EA 3900.25 CAD
100489-082 100489-080 100489-088 100489-118 100489-072 100489-090 100489-126 100489-130 100500-084 100489-078 100500-086 100489-124 100489-074 100489-120 100501-726 100489-084 100489-086 100489-114 100489-128
Lab-Tek® II Chamber Slide™ System, Electron Microscopy Sciences
Slides Chamber Slides
Lab-Tek® II Chamber Slide™ products offer optimal surface for excellent attachment and growth of cells, easily distinguished by the blue mark
  • Sterile, Chamber Slide™ products have removable, polystyrene media chamber of 1, 2, 4, and 8-well configuration
  • Low fluorescent glass microscope slide (25 x 75 x 1.2 mm) with rounded corners
  • Biocompatible adhesive
  • Polystyrene cover
  • Inert hydrophobic well border printed on slide
  • SuperFrost printed area
Lab-Tek® II CC2™ Chamber Slide™ products are the newest addition to our Lab-Tek® II Chamber Slide™ family. CC2More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable