ProSwift SCX -1S Protein Columns, Thermo Scientific
Supplier: Thermo Scientific
066765
10044-588EA 2859.38 CAD
10044-588
ProSwift SCX -1S Protein Columns, Thermo Scientific
Chromatography Columns
Fast ion-exchange separation and protein purification is possible using the Thermo Scientific™ ProSwift ion exchange (IEX) columns.
  • Durable and robust, these columns exhibit stability
  • Reproducibility even after hundreds of runs
These monolith ion-exchange columns deliver the outstanding resolving power of nonporous media combined with the high capacity of conventional porous media. They offer excellent separations using fast gradients at low and high flow rates, which improve productivity.