ProSwift RP-2H Protein Columns, Thermo Scientific
Supplier: Thermo Scientific
064296
10044-550EA 2152.06 CAD
10044-550
ProSwift RP-2H Protein Columns, Thermo Scientific
Chromatography Columns
Run protein separations at exceptionally high flow rates with Thermo Scientific™ ProSwift RP columns.
These reversed-phase monolith columns deliver the outstanding resolving power of nonporous analytical media combined with faster separations and analysis than other bead-based columns. ProSwift polymer monolith columns achieve long column lifetime, exhibit reproducibility even after hundreds of runs, and have broad pH stability, providing high stringent wash capability (1 M NaOH).
Order Now
SPECIFICATIONS
Description 10044-550
Environmentally Preferable
pH range 1 to 14