BRAND® Filtered Pipette Tips for Pipettes using 200 µl Tips, BrandTech®
Supplier: Brandtech
732728 732730 732732
10034-008PK 403.45 CAD
10034-008 10034-010 10034-012 10034-040 10034-042 10034-080
BRAND® Filtered Pipette Tips for Pipettes using 200 µl Tips, BrandTech®
Pipette Tips
BRAND filtered pipette tips for pipettes using 200 ul tips are an excellent choice to reduced contaminating aerosols when using pipettes with a yellow color code.
  • Non-sealing filters made of polyetheylene
  • Sample recovery possible if filter is wetted
  • Autoclavable at 121˚C for 20 minutes
  • Environmentally-friendly packaging
All tips are produced under cleanroom conditions. New environmentally friendly refillable TipBox design is rigid and allows partial loading of multichannels. Hinged lid can be rotated 180 degrees to be a lift-off design.More Product Information