PuriCare® Open Access Station, Labconco®
Supplier: Labconco
3820000 3820020 3820033 3820034 3820036 3820038 3820001 3820011 3820021 3820023 3820024 3820026 3820028
82003-760EA 32555.9 CAD
82003-760 82003-762 10033-454 10033-310 10033-312 10033-314 10033-316 10033-318 10033-320 10033-322 10033-324 10033-326 10033-328
PuriCare® Open Access Station, Labconco®
Fume Hoods and Enclosures Animal Research Enclosures
PuriCare Open Access Stations offer the utmost efficiency for cage changing operations
The PuriCare Open Access Station is supplied with two 99.99% efficient HEPA filters, disposable inlet and exhaust prefilters, stainless steel dished work surface, polyurethane casters, as well as ADA-compliant light and blower switches, electrical duplex, and two Minihelic® II pressure gauges. Unit features an attractive, durable glacier white and slate gray epoxy-coated steel exterior. It also features a built-in electric hydraulic lift that allows the working height to be adjusted from 93–114cm (36.5–4More Product Information