VWR® 250 Homogenizer

Supplier: VWR International
10032-338
10032-338EA 3481.67 CAD
10032-338
VWR® 250 Homogenizer
Homogenizers Handheld Homogenizers
The VWR® 250 Homogenizer unit is a perfect fit between the VWR® 200 Homogenizer unit and the benchtop VWR® Homogenizer units.

  • Versatile handheld homogenizer unit
  • Ideal for processing volumes from 0.03 ml to 5L
  • High-torque motor with variable speed adjustment of 10,000 to 30,000 rpm
  • Use handheld or post mount to a stand
  • VWR® Generator probe sold separately

This unit is lightweight enough to be handheld for convenience, yet offers extra torque for processing difficult samples

With VWR® generator probes ranging in diameter from 5mm to 30mm, it canMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR